Assay Details
| Gene Symbol: NTNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113226.2 | NP_001106697.1 | 4 - 5 | 1607 | 68 | |
| XM_011541021.1 | XP_011539323.1 | 5 - 6 | 2523 | 68 | ||
| XM_017000680.1 | XP_016856169.1 | 5 - 6 | 1889 | 68 | ||
| XM_017000681.1 | XP_016856170.1 | 5 - 6 | 2842 | 68 | ||
| GenBank mRNA | AY764260.1 | - | 2 - 3 | 51 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000926.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370076.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370076.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000462149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000462149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000462149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000462149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |

NTNG1
Genomic Map