Assay Details
| Gene Symbol: NTNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113226.2 | NP_001106697.1 | 6 - 7 | 1915 | 84 | |
| NM_001312688.1 | NP_001299617.1 | 7 - 8 | 2095 | 84 | ||
| XM_011541021.1 | XP_011539323.1 | 7 - 8 | 2831 | 84 | ||
| XM_017000680.1 | XP_016856169.1 | 7 - 8 | 2197 | 84 | ||
| XM_017000681.1 | XP_016856170.1 | 7 - 8 | 3150 | 84 | ||
| XM_017000682.1 | XP_016856171.1 | 6 - 7 | 2663 | 84 | ||
| GenBank mRNA | AK296533.1 | - | 7 - 8 | 1996 | 84 | |
| AY764260.1 | - | 4 - 5 | 359 | 84 | ||
| AY764265.1 | - | 6 - 7 | 485 | 84 | ||
| AY764266.1 | - | 4 - 5 | 257 | 84 | ||
| AY764267.1 | - | 5 - 6 | 317 | 84 | ||
| AY781194.1 | - | 3 - 4 | 191 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000926.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000294649.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370073.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370074.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370076.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000370076.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000462149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000462149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000462149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000462149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | ENST00000477948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113226.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_001113228.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009307.hg.1 | NM_014917.3 | Refseq |

NTNG1
Genomic Map