Assay Details
| Gene Symbol: AZIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301668.1 | NP_001288597.1 | 2 - 3 | 530 | 85 | |
| NM_148174.3 | NP_680479.1 | 2 - 3 | 530 | 85 | ||
| XM_005250969.1 | XP_005251026.1 | 2 - 3 | 491 | 85 | ||
| XM_011517128.2 | XP_011515430.1 | 2 - 3 | 523 | 85 | ||
| XM_017013566.1 | XP_016869055.1 | 2 - 3 | 523 | 85 | ||
| GenBank mRNA | AK095493.1 | - | 2 - 3 | 458 | 85 | |
| AU280209.1 | - | 2 - 3 | 499 | 85 | ||
| BC019279.1 | - | 2 - 3 | 477 | 2823000 | 85 | |
| BC042404.1 | - | 2 - 3 | 501 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001501.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK095493.1,BC019279.1,NM_001301668.1,NM_148174.3,XM_005250969.1,XM_011517128.2,XM_017013566.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cellular amino acid metabolic process
putrescine biosynthetic process from ornithine
negative regulation of protein catabolic process
positive regulation of catalytic activity
positive regulation of polyamine transmembrane transport
catalytic activity
protein binding
ornithine decarboxylase activator activity
regulation of cellular amino acid metabolic process
putrescine biosynthetic process from ornithine
negative regulation of protein catabolic process
positive regulation of catalytic activity
positive regulation of polyamine transmembrane transport
catalytic activity
protein binding
ornithine decarboxylase activator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000523071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000523071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000523071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |

AZIN1
Genomic Map