Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008191.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008191.hg.1 |
ENST00000421947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008191.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008191.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008191.hg.1 |
TC20000565.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000157761.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157761.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157761.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157761.hg.1 |
TC2000008198.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157761.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157761.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157761.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |