| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008190.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008190.hg.1 |
ENST00000421947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008190.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008190.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008190.hg.1 |
TC20000565.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000157760.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000157760.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000157760.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000157760.hg.1 |
TC2000008198.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000157760.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000157760.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000157760.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
ENST00000380605.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_001184699.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008198.hg.1 |
NM_019609.4 |
Refseq |