Assay Details
| Gene Symbol: CLGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130675.1 | NP_001124147.1 | 5 - 6 | 494 | 121 | |
| NM_004362.2 | NP_004353.1 | 4 - 5 | 410 | 121 | ||
| GenBank mRNA | AK093096.1 | - | 4 - 5 | 973 | 121 | |
| AK290819.1 | - | 4 - 5 | 407 | 121 | ||
| AL045602.1 | - | 5 - 6 | 438 | 121 | ||
| BC028357.2 | - | 4 - 5 | 364 | 4830135 | 121 | |
| D86322.1 | - | 4 - 5 | 377 | 121 | ||
| DC396076.1 | - | 4 - 5 | 411 | 121 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001589.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04022147.hg.1 | ENST00000325617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022147.hg.1 | ENST00000414773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022147.hg.1 | ENST00000537281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022147.hg.1 | ENST00000545667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022147.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022147.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022147.hg.1 | TC04001589.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011973.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | ENST00000325617.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | ENST00000414773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | ENST00000509477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | NM_001130675.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | NM_004362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182107.hg.1 | TC0400011973.hg.1 | Clariom D TC |

CLGN
Genomic Map