Assay Details
| Gene Symbol: ZNF329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527310.2 | XP_011525612.1 | 2 - 3 | 469 | 159 | |
| XM_011527315.1 | XP_011525617.1 | 2 - 3 | 469 | 159 | ||
| GenBank mRNA | AK090893.1 | - | 2 - 3 | 469 | 159 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK090893.1, XM_011527310.2, XM_011527315.1 transcripts.
Set Membership: |
|

ZNF329
Genomic Map