| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000369574.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000369574.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000369574.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608891.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608891.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608891.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
ENST00000608891.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000369574.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000369574.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000369574.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000388923.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608891.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608891.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608891.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
ENST00000608891.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008729.hg.1 |
NM_001010868.2 |
Refseq |