| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011211.hg.1 |
ENST00000596350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011211.hg.1 |
ENST00000596350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000600381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000600381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000600381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
ENST00000600381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011211.hg.1 |
ENST00000596350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011211.hg.1 |
ENST00000596350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000389201.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000599957.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000600381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000600381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000600381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
ENST00000600381.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011209.hg.1 |
NM_001080457.1 |
Refseq |