Assay Details
| Gene Symbol: CCDC91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005253413.1 | XP_005253470.1 | 1 - 2 | 220 | 152 | |
| XM_017019570.1 | XP_016875059.1 | 1 - 2 | 216 | 152 | ||
| XM_017019577.1 | XP_016875066.1 | 1 - 2 | 220 | 152 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005253413.1, XM_017019570.1, XM_017019577.1 transcripts.
Set Membership: |
|

CCDC91
Genomic Map