Assay Details
| Gene Symbol: TRPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130698.1 | NP_001124170.1 | 10 - 11 | 2622 | 152 | |
| NM_003305.2 | NP_003296.1 | 9 - 10 | 2404 | 152 | ||
| XM_011532217.2 | XP_011530519.1 | 11 - 12 | 2976 | 200 | ||
| GenBank mRNA | AB255423.1 | - | 8 - 9 | 2164 | 152 | |
| AB255424.1 | - | 9 - 10 | 2329 | 152 | ||
| BC093682.1 | - | 9 - 10 | 2404 | 7939527 | 152 | |
| BC093684.1 | - | 9 - 10 | 2404 | 7939529 | 152 | |
| Y13758.1 | - | 9 - 10 | 2753 | 152 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001518.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
manganese ion transport
single fertilization
phototransduction
positive regulation of calcium ion transport into cytosol
platelet activation
response to ATP
response to calcium ion
calcium ion transmembrane transport
positive regulation of cardiac muscle hypertrophy in response to stress
calcium channel activity
protein binding
store-operated calcium channel activity
inositol 1,4,5 trisphosphate binding
calcium ion transport
manganese ion transport
single fertilization
phototransduction
positive regulation of calcium ion transport into cytosol
platelet activation
response to ATP
response to calcium ion
calcium ion transmembrane transport
positive regulation of cardiac muscle hypertrophy in response to stress
calcium channel activity
protein binding
store-operated calcium channel activity
inositol 1,4,5 trisphosphate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000502968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000502968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021577.hg.1 | ENST00000264811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021577.hg.1 | ENST00000379645.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021577.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021577.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021577.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021577.hg.1 | TC04001518.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000502968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000502968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011750.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | ENST00000264811.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | ENST00000379645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | ENST00000506449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | ENST00000513531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | NM_001130698.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | NM_003305.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180872.hg.1 | TC0400011750.hg.1 | Clariom D TC |

TRPC3
Genomic Map