| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000506508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000506508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000506508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000509419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000509419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
ENST00000509419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013723.hg.1 |
ENST00000307746.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013723.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013723.hg.1 |
TC04000993.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400167930.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167930.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167930.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167930.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167930.hg.1 |
TC0400009879.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000307746.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000506508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000506508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000506508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000509419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000509419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
ENST00000509419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009879.hg.1 |
NM_018659.2 |
Refseq |