Assay Details
| Gene Symbol: NOC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022451.10 | NP_071896.8 | 2 - 3 | 348 | 81 | |
| GenBank mRNA | AB077992.1 | - | 2 - 3 | 235 | 81 | |
| AK091246.1 | - | 2 - 3 | 305 | 81 | ||
| AK292520.1 | - | 2 - 3 | 330 | 81 | ||
| BC049850.1 | - | 2 - 3 | 298 | 5549517 | 81 | |
| BX505710.1 | - | 2 - 3 | 350 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001545.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10019435.hg.1 | ENST00000371350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019435.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019435.hg.1 | ENST00000463649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019435.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019435.hg.1 | TC10001545.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019437.hg.1 | ENST00000371350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019437.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019437.hg.1 | ENST00000463649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019437.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019437.hg.1 | TC10001545.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019439.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019439.hg.1 | ENST00000463649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019439.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019439.hg.1 | TC10001545.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000177887.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177887.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177887.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000177887.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177887.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177887.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000177887.hg.1 | TC1000011469.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000177889.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177889.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177889.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177889.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000177889.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177889.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177889.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177889.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000177889.hg.1 | TC1000011469.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000177891.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177891.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177891.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177891.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000177891.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177891.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177891.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177891.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000177891.hg.1 | TC1000011469.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 185299 | - | - |

NOC3L
Genomic Map