Assay Details
Gene Symbol: NOC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022451.10 | NP_071896.8 | 3 - 4 | 486 | 150 | |
GenBank mRNA | AB077992.1 | - | 3 - 4 | 373 | 150 | |
AK091246.1 | - | 3 - 4 | 443 | 150 | ||
AK292520.1 | - | 3 - 4 | 468 | 150 | ||
BC049850.1 | - | 3 - 4 | 436 | 5549517 | 150 | |
BX505710.1 | - | 3 - 4 | 488 | 150 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001545.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10019432.hg.1 | ENST00000371350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10019432.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10019432.hg.1 | ENST00000463649.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10019432.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10019432.hg.1 | TC10001545.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10019431.hg.1 | ENST00000371350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10019431.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10019431.hg.1 | ENST00000463649.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10019431.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10019431.hg.1 | TC10001545.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10019433.hg.1 | ENST00000371350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10019433.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10019433.hg.1 | ENST00000463649.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10019433.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10019433.hg.1 | TC10001545.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177884.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177884.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177884.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177884.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177884.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177884.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177884.hg.1 | TC1000011469.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000177885.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177885.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177885.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177885.hg.1 | ENST00000371361.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177885.hg.1 | ENST00000463649.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000177885.hg.1 | NM_022451.10 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000177885.hg.1 | TC1000011469.hg.1 | Clariom D TC |
