Assay Details
| Gene Symbol: NOC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022451.10 | NP_071896.8 | 9 - 10 | 1269 | 147 | |
| XM_017016544.1 | XP_016872033.1 | 7 - 8 | 1322 | 147 | ||
| GenBank mRNA | AK022882.1 | - | 1 - 2 | 180 | 147 | |
| AK091246.1 | - | 9 - 10 | 1226 | 147 | ||
| AK292520.1 | - | 9 - 10 | 1251 | 147 | ||
| AK302028.1 | - | 3 - 4 | 354 | 147 | ||
| BC049850.1 | - | 9 - 10 | 1219 | 5549517 | 147 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001545.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK022882.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019426.hg.1 | ENST00000371350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019426.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019426.hg.1 | ENST00000463649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019426.hg.1 | ENST00000543788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019426.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10019426.hg.1 | TC10001545.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000177879.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177879.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177879.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000177879.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177879.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000177879.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000177879.hg.1 | TC1000011469.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000371361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000461562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | ENST00000463649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011469.hg.1 | NM_022451.10 | Refseq |

NOC3L
Genomic Map