| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525499.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525499.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000525662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027306.hg.1 |
ENST00000393067.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027306.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027306.hg.1 |
TC11002278.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100205338.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100205338.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100205338.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100205338.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100205338.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100205338.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100205338.hg.1 |
TC1100013224.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000393067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525499.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525499.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525584.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000525662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000526535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
ENST00000530793.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013224.hg.1 |
NM_006235.2 |
Refseq |