Assay Details
Gene Symbol: RPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000981.3 | NP_000972.1 | 1 - 2 | 67 | 142 | |
GenBank mRNA | BC095445.1 | - | 1 - 2 | 67 | 30721753 | 142 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000473.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC095445.1,NM_000981.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500012801.hg.1 | ENST00000524262.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007570.hg.1 | ENST00000438827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17005538.hg.1 | ENST00000225430.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17005538.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005538.hg.1 | TC17000473.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000577759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005539.hg.1 | ENST00000225430.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17005539.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005539.hg.1 | TC17000473.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17005533.hg.1 | ENST00000225430.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17005533.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005533.hg.1 | TC17000473.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155849.hg.1 | TC1700007740.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300007570.hg.1 | ENST00000438827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155835.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155835.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155835.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155835.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155835.hg.1 | TC1700007740.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155847.hg.1 | TC1700007740.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500012801.hg.1 | ENST00000524262.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000225430.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000577759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000579374.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000580333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000582193.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | ENST00000585199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007740.hg.1 | NM_000981.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 133819 | - | - |
