Assay Details
| Gene Symbol: NEGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173808.2 | NP_776169.2 | 4 - 5 | 772 | 158 | |
| XM_011541200.2 | XP_011539502.1 | 4 - 5 | 1028 | 158 | ||
| GenBank mRNA | AK092307.1 | - | 4 - 5 | 679 | 158 | |
| BC036771.1 | - | 4 - 5 | 694 | 5576828 | 158 | |
| BX538014.1 | - | 4 - 5 | 676 | 158 | ||
More Information
Set Membership: |
|

NEGR1
Genomic Map