Assay Details
Gene Symbol: LOC730961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC034282.1 | - | 1 | 1164 | 128 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein phosphatase 2 catalytic subunit alpha | PPP2CA |
PP2Ac, PP2CA, PP2Calpha, RP-C |
-884.0 | Anti-sense | Chr. 5 - 134196457 - 134226259 on NCBI Build GRCh38 |
Downstream | cyclin dependent kinase like 3 | CDKL3 | NKIAMRE | 8.4kb | Anti-sense | Chr. 5 - 134298424 - 134371159 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
