| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000406880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000406880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000406880.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007817.hg.1 |
ENST00000358295.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007817.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007817.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22007817.hg.1 |
TC22000407.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2200169462.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169462.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200169462.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200169462.hg.1 |
TC2200009290.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200169462.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200169462.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200169462.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000358295.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000402357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000406880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000406880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000406880.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
ENST00000473898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_001082967.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009290.hg.1 |
NM_015381.6 |
Refseq |