Assay Details
| Gene Symbol: PCED1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281429.1 | NP_001268358.1 | 1 - 2 | 119 | 159 | |
| NM_138371.2 | NP_612380.1 | 1 - 2 | 119 | 159 | ||
| GenBank mRNA | BC072670.1 | - | 1 - 2 | 119 | 159 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000334.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001281429.1,NM_138371.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000548348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000548348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000548348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000551777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000551777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000551777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | ENST00000551777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004221.hg.1 | ENST00000432328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004221.hg.1 | ENST00000546455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004221.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004221.hg.1 | TC12000334.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200152445.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152445.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152445.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152445.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152445.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152445.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152445.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152445.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152445.hg.1 | TC1200007461.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200152447.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152447.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152447.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152447.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152447.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152447.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200152447.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152447.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200152447.hg.1 | TC1200007461.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000432328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000546455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000548348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000548348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000548348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549500.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000549630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000551777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000551777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000551777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | ENST00000551777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_001281429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007461.hg.1 | NM_138371.2 | Refseq |

PCED1B
Genomic Map