| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000452236.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000526291.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
ENST00000526291.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014250.hg.1 |
NM_052923.1 |
Refseq |