Assay Details
| Gene Symbol: ZNF329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095349.1 | - | 2 - 3 | 129 | 168 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 195631 | - | - |

ZNF329
Genomic Map