Assay Details
| Gene Symbol: TMEM132D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133448.2 | NP_597705.2 | 6 - 7 | 1977 | 82 | |
| GenBank mRNA | AB061814.1 | - | 6 - 7 | 1978 | 82 | |
| AB075824.1 | - | 1 - 2 | 1504 | 82 | ||
| BC114626.1 | - | 6 - 7 | 1977 | 40082027 | 82 | |
| BC114936.1 | - | 6 - 7 | 1977 | 40082028 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002124.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028002.hg.1 | ENST00000389441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028002.hg.1 | ENST00000422113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028002.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028002.hg.1 | TC12002124.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028006.hg.1 | ENST00000422113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028006.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028006.hg.1 | TC12002124.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028008.hg.1 | ENST00000422113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028008.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028008.hg.1 | TC12002124.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028004.hg.1 | ENST00000422113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028004.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028004.hg.1 | TC12002124.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028007.hg.1 | ENST00000422113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028007.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028007.hg.1 | TC12002124.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028001.hg.1 | ENST00000389441.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028001.hg.1 | ENST00000422113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028001.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028001.hg.1 | TC12002124.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028005.hg.1 | ENST00000422113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028005.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028005.hg.1 | TC12002124.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200197215.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197215.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197215.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197215.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197215.hg.1 | TC1200012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200197206.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197206.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197206.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197206.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197206.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197206.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197206.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197206.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197206.hg.1 | TC1200012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200197207.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197207.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197207.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197207.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197207.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197207.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197207.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197207.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197207.hg.1 | TC1200012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200197216.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197216.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197216.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197216.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197216.hg.1 | TC1200012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200197211.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197211.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197211.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197211.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197211.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197211.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197211.hg.1 | TC1200012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200197213.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197213.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197213.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197213.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197213.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197213.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197213.hg.1 | TC1200012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200197210.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197210.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197210.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197210.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197210.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197210.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197210.hg.1 | TC1200012424.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000389441.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012424.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197209.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197209.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197209.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197209.hg.1 | ENST00000422113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197209.hg.1 | ENST00000619366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197209.hg.1 | NM_133448.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197209.hg.1 | TC1200012424.hg.1 | Clariom D TC |

TMEM132D
Genomic Map