Assay Details
| Gene Symbol: SASS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304829.1 | NP_001291758.1 | 6 - 7 | 875 | 126 | |
| NM_194292.2 | NP_919268.1 | 7 - 8 | 955 | 126 | ||
| XM_017000486.1 | XP_016855975.1 | 7 - 8 | 721 | 126 | ||
| GenBank mRNA | AK302513.1 | - | 8 - 9 | 705 | 126 | |
| AK307824.1 | - | 6 - 7 | 742 | 126 | ||
| AL834265.1 | - | 3 - 4 | 218 | 126 | ||
| AY359522.1 | - | 7 - 8 | 815 | 126 | ||
| BC101026.3 | - | 6 - 7 | 668 | 40007432 | 126 | |
| BX641109.1 | - | 7 - 8 | 774 | 126 | ||
| BX648406.1 | - | 7 - 8 | 126 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002916.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045315.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045315.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045315.hg.1 | ENST00000535161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045315.hg.1 | ENST00000539329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045315.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045315.hg.1 | TC01002916.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212168.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212168.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212168.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212168.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212168.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212168.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212168.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212168.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212168.hg.1 | TC0100015058.hg.1 | Clariom D TC |

SASS6
Genomic Map