Assay Details
| Gene Symbol: GLIS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147193.2 | NP_671726.2 | 1 - 2 | 306 | 76 | |
| GenBank mRNA | AK090634.1 | - | 1 - 2 | 305 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002674.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK090634.1,NM_147193.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041974.hg.1 | ENST00000312233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041974.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041974.hg.1 | TC01002674.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041975.hg.1 | ENST00000312233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041975.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041975.hg.1 | TC01002674.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100206622.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206622.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206622.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206622.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206622.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206622.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206622.hg.1 | TC0100014240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100206623.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206623.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206623.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100206623.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100206623.hg.1 | TC0100014240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000312233.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | ENST00000628545.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014240.hg.1 | NM_147193.2 | Refseq |

GLIS1
Genomic Map