| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030220.hg.1 |
ENST00000263989.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030220.hg.1 |
ENST00000398569.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030220.hg.1 |
ENST00000398571.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030220.hg.1 |
NM_014709.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030220.hg.1 |
TC02001891.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200192904.hg.1 |
ENST00000398571.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192904.hg.1 |
NM_014709.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192904.hg.1 |
ENST00000398571.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200192904.hg.1 |
NM_014709.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200192904.hg.1 |
TC0200012734.hg.1 |
Clariom D TC |