Assay Details
| Gene Symbol: ZNF329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527316.2 | XP_011525618.1 | 1 - 2 | 57 | 76 | |
| GenBank mRNA | AK096542.1 | - | 1 - 2 | 38 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK096542.1, XM_011527316.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000358067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000500161.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000597186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000598312.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | ENST00000601887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011606.hg.1 | NM_024620.3 | Refseq |

ZNF329
Genomic Map