Assay Details
| Gene Symbol: UBXN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077619.1 | NP_001071087.1 | 1 - 2 | 204 | 137 | |
| XM_005251165.2 | XP_005251222.1 | 1 - 2 | 107 | 137 | ||
| XM_005251166.1 | XP_005251223.1 | 1 - 2 | 107 | 137 | ||
| GenBank mRNA | AK126300.1 | - | 1 - 2 | 107 | 137 | |
| AL831990.2 | - | 1 - 2 | 78 | 137 | ||
| AL833075.2 | - | 1 - 2 | 78 | 137 | ||
| BC113645.1 | - | 1 - 2 | 204 | 8322697 | 137 | |
| BX647876.2 | - | 1 - 2 | 78 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000402.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005621.hg.1 | ENST00000399598.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005621.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005621.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005621.hg.1 | TC08000402.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005618.hg.1 | ENST00000399598.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005618.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005618.hg.1 | TC08000402.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005623.hg.1 | ENST00000399598.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005623.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005623.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005623.hg.1 | TC08000402.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013241.hg.1 | ENST00000451057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013241.hg.1 | ENST00000451057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153220.hg.1 | TC0800007735.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013241.hg.1 | ENST00000451057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013241.hg.1 | ENST00000451057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153222.hg.1 | TC0800007735.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800153217.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153217.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153217.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153217.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153217.hg.1 | TC0800007735.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 175389 | - | - |

UBXN2B
Genomic Map