Assay Details
| Gene Symbol: UBXN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077619.1 | NP_001071087.1 | 2 - 3 | 310 | 165 | |
| XM_005251165.2 | XP_005251222.1 | 2 - 3 | 213 | 165 | ||
| XM_005251166.1 | XP_005251223.1 | 2 - 3 | 213 | 165 | ||
| GenBank mRNA | AK126300.1 | - | 2 - 3 | 213 | 165 | |
| AL831990.2 | - | 2 - 3 | 184 | 165 | ||
| AL833075.2 | - | 2 - 3 | 184 | 165 | ||
| BC113645.1 | - | 2 - 3 | 310 | 8322697 | 165 | |
| BX647876.2 | - | 2 - 3 | 184 | 165 | ||
More Information
Set Membership: |
|

UBXN2B
Genomic Map