Assay Details
| Gene Symbol: GRAMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146319.1 | NP_001139791.1 | 13 - 14 | 1531 | 150 | |
| NM_001146320.1 | NP_001139792.1 | 12 - 13 | 1568 | 150 | ||
| NM_001146321.1 | NP_001139793.1 | 14 - 15 | 1723 | 150 | ||
| NM_001146322.1 | NP_001139794.1 | 13 - 14 | 1342 | 150 | ||
| NM_023927.2 | NP_076416.2 | 13 - 14 | 1680 | 150 | ||
| XM_005272057.3 | XP_005272114.1 | 13 - 14 | 1421 | 150 | ||
| XM_005272058.3 | XP_005272115.1 | 13 - 14 | 1418 | 150 | ||
| XM_005272059.4 | XP_005272116.1 | 14 - 15 | 1811 | 150 | ||
| XM_005272060.3 | XP_005272117.1 | 13 - 14 | 1508 | 150 | ||
| XM_005272061.3 | XP_005272118.1 | 13 - 14 | 1503 | 150 | ||
| XM_005272062.3 | XP_005272119.1 | 13 - 14 | 1469 | 150 | ||
| XM_005272063.3 | XP_005272120.1 | 13 - 14 | 1615 | 150 | ||
| XM_005272064.3 | XP_005272121.1 | 14 - 15 | 1528 | 150 | ||
| XM_011543591.2 | XP_011541893.1 | 14 - 15 | 2399 | 150 | ||
| XM_011543592.2 | XP_011541894.1 | 14 - 15 | 2392 | 150 | ||
| XM_011543593.2 | XP_011541895.1 | 13 - 14 | 1508 | 150 | ||
| XM_011543594.2 | XP_011541896.1 | 13 - 14 | 1280 | 150 | ||
| XM_011543595.2 | XP_011541897.1 | 12 - 13 | 1275 | 150 | ||
| XM_017009781.1 | XP_016865270.1 | 14 - 15 | 1286 | 150 | ||
| XM_017009782.1 | XP_016865271.1 | 14 - 15 | 1525 | 150 | ||
| XM_017009784.1 | XP_016865273.1 | 12 - 13 | 1327 | 150 | ||
| XM_017009785.1 | XP_016865274.1 | 12 - 13 | 1377 | 150 | ||
| GenBank mRNA | AK024966.1 | - | 13 - 14 | 1448 | 150 | |
| AK225829.1 | - | 13 - 14 | 1448 | 150 | ||
| AK293307.1 | - | 12 - 13 | 1273 | 150 | ||
| AK296267.1 | - | 14 - 15 | 1428 | 150 | ||
| AK300154.1 | - | 13 - 14 | 1476 | 150 | ||
| AK303857.1 | - | 13 - 14 | 1282 | 150 | ||
| AK316288.1 | - | 13 - 14 | 1495 | 150 | ||
| BC008590.1 | - | 13 - 14 | 1413 | 4148772 | 150 | |
More Information
Set Membership: |
|

GRAMD3
Genomic Map