Assay Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173564.3 | NP_775835.2 | 5 - 6 | 2318 | 104 | |
| XM_006715907.3 | XP_006715970.1 | 5 - 6 | 2376 | 104 | ||
| GenBank mRNA | AK094857.1 | - | 2 - 3 | 429 | 104 | |
| AK124773.1 | - | 5 - 6 | 2318 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000623.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000489641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000489641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000489641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009497.hg.1 | ENST00000300179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009497.hg.1 | ENST00000423930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009497.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009497.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009497.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009497.hg.1 | TC07000623.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009499.hg.1 | ENST00000300179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009499.hg.1 | ENST00000423930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009499.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009499.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009499.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009499.hg.1 | TC07000623.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | ENST00000489641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | ENST00000489641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700160161.hg.1 | TC0700008548.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000489641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000489641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000489641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008548.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700160159.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160159.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160159.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160159.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700160159.hg.1 | ENST00000300179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160159.hg.1 | ENST00000454988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160159.hg.1 | ENST00000496985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160159.hg.1 | NM_173564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700160159.hg.1 | TC0700008548.hg.1 | Clariom D TC |

NYAP1
Genomic Map