Assay Details
| Gene Symbol: TDG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003211.4 | NP_003202.3 | 1 | 65 | 121 | |
| GenBank mRNA | AL710869.1 | - | 1 | 59 | 121 | |
| DB162106.1 | - | 1 | 65 | 121 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000804.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_003211.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
base-excision repair
base-excision repair, AP site formation
mismatch repair
transcription, DNA-templated
embryo development
covalent chromatin modification
protein sumoylation
negative regulation of protein binding
oxidative DNA demethylation
negative regulation of chromatin binding
regulation of gene expression, epigenetic
depyrimidination
DNA demethylation
RNA polymerase II transcription cofactor activity
damaged DNA binding
double-stranded DNA binding
uracil DNA N-glycosylase activity
protein kinase C binding
protein binding
pyrimidine-specific mismatch base pair DNA N-glycosylase activity
DNA N-glycosylase activity
mismatched DNA binding
SUMO binding
protein homodimerization activity
structure-specific DNA binding
negative regulation of transcription from RNA polymerase II promoter
base-excision repair
base-excision repair, AP site formation
mismatch repair
transcription, DNA-templated
embryo development
covalent chromatin modification
protein sumoylation
negative regulation of protein binding
oxidative DNA demethylation
negative regulation of chromatin binding
regulation of gene expression, epigenetic
depyrimidination
DNA demethylation
RNA polymerase II transcription cofactor activity
damaged DNA binding
double-stranded DNA binding
uracil DNA N-glycosylase activity
protein kinase C binding
protein binding
pyrimidine-specific mismatch base pair DNA N-glycosylase activity
DNA N-glycosylase activity
mismatched DNA binding
SUMO binding
protein homodimerization activity
structure-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007115.hg.1 | ENST00000543961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000266775.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000392872.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000401157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000436021.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000436021.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000436021.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000436021.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000436021.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000436021.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000436021.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000536395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000536395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000536395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000537100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000537100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000537100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000537100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000537100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000537100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000537100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000537100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000537100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000540956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000542926.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000544861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000545698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000545698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000545698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000545698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | ENST00000545698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008669.hg.1 | NM_003211.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007115.hg.1 | ENST00000543961.1 | Ensembl |

TDG
Genomic Map