| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002085.hg.1 |
ENST00000354348.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002085.hg.1 |
NM_003495.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002085.hg.1 |
TC06000224.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014084.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014084.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014084.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014084.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014084.hg.1 |
NM_003495.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014084.hg.1 |
NM_003495.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197772.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197772.hg.1 |
NM_003495.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197772.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600197772.hg.1 |
NM_003495.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600197772.hg.1 |
TC0600014084.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600014084.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014084.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014084.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014084.hg.1 |
ENST00000615353.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014084.hg.1 |
NM_003495.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014084.hg.1 |
NM_003495.2 |
Refseq |