| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014300.hg.1 |
ENST00000357802.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014300.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014300.hg.1 |
ENST00000437211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014300.hg.1 |
ENST00000458605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014300.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014300.hg.1 |
TC22000847.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014301.hg.1 |
ENST00000357802.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014301.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014301.hg.1 |
ENST00000437211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014301.hg.1 |
ENST00000458605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014301.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014301.hg.1 |
TC22000847.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000516104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
TC2200008883.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164363.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000516104.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008883.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
NR_002184.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
TC2200008883.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000357802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000421116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000437211.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000458605.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
ENST00000566851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200164364.hg.1 |
NR_002184.2 |
Refseq |