Assay Details
| Gene Symbol: TOP1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002719.2 | 1 | 449 | 161 | ||
| Gene Symbol: FMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011509347.2 | XP_011507649.1 | 10 | 1802 | 161 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | flavin containing monooxygenase 1 | FMO1 | -52.9kb | Sense | Chr. 1 - 171248471 - 171285978 on NCBI Build GRCh38 | |
| Downstream | proline rich coiled-coil 2C | PRRC2C |
BAT2-iso, BAT2D1, BAT2L2, XTP2 |
144.2kb | Sense | Chr. 1 - 171485496 - 171593511 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3841 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P305507 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594221 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15143312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653636 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653683 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653654 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653649 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594221 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15143312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653636 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653647 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653683 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653657 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00594247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20653668 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15143314 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594197 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594208 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594190 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594182 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594238 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594224 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594185 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594250 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594219 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594222 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594230 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594209 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594216 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594187 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594203 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594236 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594193 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594213 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15143318 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594247 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15143313 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15143320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594245 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594201 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594205 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15143334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15143316 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594195 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594248 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594241 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00594226 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034907 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034907 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 115448 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1802922 | NR_002719.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1802922 | M55632.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2086952 | M55632.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1802922 | NR_002719.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1802922 | M55632.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1802922 | NR_002719.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1802922 | M55632.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2086952 | M55632.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2086952 | M55632.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2086952 | M55632.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1802922 | NR_002719.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1802922 | M55632.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2086952 | M55632.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2086952 | M55632.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1802922 | NR_002719.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1802922 | M55632.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1802922 | NR_002719.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1802922 | M55632.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2086952 | M55632.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1802922 | NR_002719.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1802922 | M55632.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2086952 | M55632.1 | human_transcriptome |

TOP1P1
Genomic Map