Assay Details
| Gene Symbol: GLYATL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010904.1 | NP_001010904.1 | 2 - 3 | 198 | 111 | |
| GenBank mRNA | DB246049.1 | - | 2 - 3 | 198 | 111 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000641.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007790.hg.1 | ENST00000371197.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007790.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007790.hg.1 | TC06000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007791.hg.1 | ENST00000371197.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007791.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007791.hg.1 | TC06000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007792.hg.1 | ENST00000371197.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007792.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007792.hg.1 | TC06000641.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600157434.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157434.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157434.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157434.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157434.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157434.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157434.hg.1 | TC0600008211.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008211.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157433.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157433.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157433.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157433.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157433.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157433.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157433.hg.1 | TC0600008211.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600157432.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157432.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157432.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157432.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157432.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157432.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157432.hg.1 | TC0600008211.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600157430.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157430.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157430.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157430.hg.1 | ENST00000371197.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157430.hg.1 | ENST00000545705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600157430.hg.1 | NM_001010904.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600157430.hg.1 | TC0600008211.hg.1 | Clariom D TC |

GLYATL3
Genomic Map