Assay Details
| Gene Symbol: VN1R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173857.2 | NP_776256.2 | 1 | 753 | 72 | |
| XM_011526885.1 | XP_011525187.1 | 2 | 905 | 72 | ||
| GenBank mRNA | BC104935.1 | - | 1 | 807 | 8143938 | 72 |
| BC104939.1 | - | 1 | 811 | 8143942 | 72 | |
More Information
Set Membership: |
|

VN1R4
Genomic Map