| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010699.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010699.hg.1 |
TC04000775.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010697.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010697.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010697.hg.1 |
TC04000775.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010698.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010698.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010698.hg.1 |
TC04000775.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400162114.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162114.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162114.hg.1 |
TC0400009048.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009048.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162113.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162113.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162113.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162113.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162113.hg.1 |
TC0400009048.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400162112.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162112.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162112.hg.1 |
ENST00000260010.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162112.hg.1 |
NM_003264.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162112.hg.1 |
TC0400009048.hg.1 |
Clariom D TC |