Assay Details
| Gene Symbol: LOC100132831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027761.1 | 1 | 716 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000968.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P41496 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 37258 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48186 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P41496 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P41496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715969 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715974 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421699 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715978 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421687 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715984 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421688 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421668 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421687 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715969 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715974 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715978 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715984 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421699 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17336222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11715979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32421688 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17336229 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715986 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715962 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17336219 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715971 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715960 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715976 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715985 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715978 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715957 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715975 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715967 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715969 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715982 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17336217 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715965 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715981 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715983 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715973 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11715980 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03694427 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012943.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012943.hg.1 | TC0X000968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164638.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164638.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164638.hg.1 | TC0X00009445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00164635.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00164635.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164635.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00164635.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164635.hg.1 | TC0X00009445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00164639.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164639.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164639.hg.1 | TC0X00009445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00164633.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164633.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164633.hg.1 | TC0X00009445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00164636.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00164636.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164636.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00164636.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164636.hg.1 | TC0X00009445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00164637.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00164637.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164637.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00164637.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164637.hg.1 | TC0X00009445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009445.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164634.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00164634.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164634.hg.1 | ENST00000424288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00164634.hg.1 | NR_027761.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00164634.hg.1 | TC0X00009445.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039762 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039763 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039762 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039763 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 230852 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3249275 | NR_027761.1 | human_transcriptome |

LOC100132831
Genomic Map