Assay Details
| Gene Symbol: MORN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538214.2 | XP_011536516.1 | 1 | 67 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011538214.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000355329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000392462.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | ENST00000542364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | NM_173855.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012210.hg.1 | NM_173855.4 | Refseq |

MORN3
Genomic Map