Assay Details
| Gene Symbol: HIST1H2AM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003514.2 | NP_003505.1 | 1 | 158 | 123 | |
| GenBank mRNA | BC032686.1 | - | 1 | 173 | 123 | |
| BC032756.1 | - | 1 | 173 | 123 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001430.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820577 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 133790 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 151 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P86389 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P221799 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P221799 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P86389 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 8138 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 17913 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P110602 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P110602 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 61873 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 5293 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 41462 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P86389 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P221799 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P86389 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P221799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630371 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P86389 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P221799 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P110602 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630373 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04630370 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04630371 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P110602 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P110602 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P221799 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P86389 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 6943 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 2391 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 27403 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P110602 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P221799 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630354 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630370 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630361 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630352 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630348 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630359 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630351 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630357 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P110602 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_003514_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8124545 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8124546 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8124541 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8124543 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8124544 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8124542 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8124540 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 214481_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11740125_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11740125_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11740124_at | - | - |
| Affymetrix Human Genome U219 Array | 11740124_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007383.hg.1 | ENST00000627103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016372.hg.1 | ENST00000359611.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016372.hg.1 | NM_003514.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06016372.hg.1 | TC06001430.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600175746.hg.1 | NM_003514.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600175746.hg.1 | NM_003514.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600175746.hg.1 | TC0600011235.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007383.hg.1 | ENST00000627103.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 214481_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208583_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207156_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214542_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207156_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214554_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214542_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214554_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214481_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208583_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 208583_x_at | - | - |
| Human Genome U133A 2.0 Array | 208583_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 214481_PM_at | - | - |
| Human Genome U133A 2.0 Array | 214554_at | - | - |
| Human Genome U133A 2.0 Array | 214554_PM_at | - | - |
| Human Genome U133A 2.0 Array | 214542_x_at | - | - |
| Human Genome U133A 2.0 Array | 214542_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 207156_PM_at | - | - |
| Human Genome U133A 2.0 Array | 207156_at | - | - |
| Human Genome U133A 2.0 Array | 214481_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003514_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw567562 | - | - |
| Human Genome U95 Set | 284_at | - | - |
| Human Genome U95 Set | 34157_PM_f_at | - | - |
| Human Genome U95 Set | 285_PM_g_at | - | - |
| Human Genome U95 Set | 34156_i_at | - | - |
| Human Genome U95 Set | 285_g_at | - | - |
| Human Genome U95 Set | 33023_at | - | - |
| Human Genome U95 Set | 34157_f_at | - | - |
| Human Genome U95 Set | 33023_PM_at | - | - |
| Human Genome U95 Set | 31768_at | - | - |
| Human Genome U95 Set | 34156_PM_i_at | - | - |
| Human Genome U95 Set | 31768_PM_at | - | - |
| Human Genome U95 Set | 284_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214481_at3 | - | - |
| Human Genome U133 Set | 208583_PM_x_at | - | - |
| Human Genome U133 Set | 207156_at | - | - |
| Human Genome U133 Set | 214542_x_at | - | - |
| Human Genome U133 Set | 214554_at | - | - |
| Human Genome U133 Set | 207156_PM_at | - | - |
| Human Genome U133 Set | 214554_PM_at | - | - |
| Human Genome U133 Set | 214481_at | - | - |
| Human Genome U133 Set | 214542_PM_x_at | - | - |
| Human Genome U133 Set | 208583_x_at | - | - |
| Human Genome U133 Set | 214481_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029577 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029577 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 179147 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_15718718-S | BC032756.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_15718718-S | NM_003514.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_26622-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0005340519 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0005340519 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 6060193 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_26622 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1756022 | NM_003514.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1756022 | BC032756.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1756022 | NM_003514.2 | human_transcriptome |

HIST1H2AM
Genomic Map