Assay Details
| Gene Symbol: CDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004065.2 | NP_004056.2 | 1 | 366 | 135 | |
| GenBank mRNA | BC113472.1 | - | 1 | 291 | 8322524 | 135 |
| BC113474.1 | - | 1 | 291 | 8322526 | 135 | |
| M16965.1 | - | 1 | 234 | 135 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CDR1
Genomic Map