Assay Details
| Gene Symbol: OR5H6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005479.1 | NP_001005479.1 | 1 | 75 | 155 | |
| GenBank mRNA | BC137523.1 | - | 1 | 116 | 9021523 | 155 |
| BC137524.1 | - | 1 | 116 | 9021524 | 155 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000487.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02438894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02438901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577238 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577217 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577226 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02438886 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15467823 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02438899 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02438895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15467831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577216 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577207 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577220 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577224 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02438888 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577223 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15467820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577212 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02438905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02438907 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02438887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22577202 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3318653 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0661 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24370 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00766743 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3318653 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3318653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15467826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438894 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15467823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438905 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15467823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438894 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02438905 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15467826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22577207 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438892 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438887 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438894 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438889 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438885 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438897 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438908 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438902 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438896 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438900 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438904 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438907 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02438906 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438900 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438892 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438896 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438910 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15467819 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15467835 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438894 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438887 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438907 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438905 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438889 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438912 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02438904 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001005479_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8081199 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8081203 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8081202 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8081198 | - | - |
| Affymetrix Human Genome U219 Array | 11738692_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738692_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008104.hg.1 | ENST00000514084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008101.hg.1 | ENST00000502817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008099.hg.1 | ENST00000356526.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008099.hg.1 | ENST00000356526.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008099.hg.1 | ENST00000503164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008099.hg.1 | ENST00000503164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008099.hg.1 | ENST00000615035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008099.hg.1 | ENST00000615035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008099.hg.1 | NM_001005515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008099.hg.1 | NM_001005515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008098.hg.1 | ENST00000437310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008098.hg.1 | ENST00000437310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008098.hg.1 | ENST00000437310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008098.hg.1 | NM_001005514.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008102.hg.1 | ENST00000510851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008105.hg.1 | ENST00000383696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008105.hg.1 | ENST00000623332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008105.hg.1 | ENST00000623942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008105.hg.1 | NM_001005479.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009311.hg.1 | ENST00000383696.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009311.hg.1 | NM_001005479.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009311.hg.1 | TC03000487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008103.hg.1 | ENST00000394191.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008103.hg.1 | ENST00000394191.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008103.hg.1 | ENST00000394191.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008103.hg.1 | ENST00000394191.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008103.hg.1 | ENST00000508616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008102.hg.1 | ENST00000510851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157742.hg.1 | TC0300008105.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008101.hg.1 | ENST00000502817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008105.hg.1 | ENST00000383696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008105.hg.1 | ENST00000623332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008105.hg.1 | ENST00000623942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008105.hg.1 | NM_001005479.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008098.hg.1 | ENST00000437310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008098.hg.1 | ENST00000437310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008098.hg.1 | ENST00000437310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008098.hg.1 | NM_001005514.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008099.hg.1 | ENST00000356526.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008099.hg.1 | ENST00000356526.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008099.hg.1 | ENST00000503164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008099.hg.1 | ENST00000503164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008099.hg.1 | ENST00000615035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008099.hg.1 | ENST00000615035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008099.hg.1 | NM_001005515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008099.hg.1 | NM_001005515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157743.hg.1 | ENST00000383696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157743.hg.1 | ENST00000623332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157743.hg.1 | ENST00000623942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157743.hg.1 | NM_001005479.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157743.hg.1 | ENST00000383696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157743.hg.1 | ENST00000623332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157743.hg.1 | ENST00000623942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157743.hg.1 | NM_001005479.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157743.hg.1 | TC0300008105.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008104.hg.1 | ENST00000514084.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022900 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022900 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_13112-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 5490402 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_13112 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1766438 | NM_001005479.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1766438 | BC137523.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1766438 | BC137524.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1766438 | NM_001005479.1 | human_transcriptome |

OR5H6
Genomic Map