Assay Details
| Gene Symbol: OR8K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002907.1 | NP_001002907.1 | 1 | 741 | 134 | |
| GenBank mRNA | BC146844.1 | - | 1 | 835 | 9056772 | 134 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000451.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16453300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218436 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218431 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218426 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218421 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218391 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218437 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218403 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218425 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218423 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218435 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218409 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218418 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218433 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218393 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218416 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218424 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218432 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218419 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218427 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218417 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218428 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218399 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218413 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218411 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218420 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218408 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808903 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218406 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 241211 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3305373 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0241 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0239 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0240 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4773 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P32910162 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3305373 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3305373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808931 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218419 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218403 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218424 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218417 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218413 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218423 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218409 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218425 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218418 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218406 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808921 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808905 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808917 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218411 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218421 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218425 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218417 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218424 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808931 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218409 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218411 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218421 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808921 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218413 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218419 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808905 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808917 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218423 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218403 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218418 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218406 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218395 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P19245401 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808904 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808930 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808910 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808932 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808901 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808908 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808912 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808899 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808915 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808919 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808926 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808917 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808906 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808921 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808903 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808934 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 38109 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 117983 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 54908 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32910162 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808896 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808912 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808903 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808936 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808940 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808979 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808910 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808963 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808960 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808934 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808915 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808930 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808978 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808981 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808906 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808976 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808917 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808904 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808972 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808965 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808919 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808943 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808970 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808898 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808901 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808983 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808968 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808921 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808932 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808946 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808926 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808908 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808949 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16453310 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808986 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808938 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001002907_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7939980 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7939979 | - | - |
| Affymetrix Human Genome U219 Array | 11742341_at | - | - |
| Affymetrix Human Genome U219 Array | 11742341_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007634.hg.1 | ENST00000279783.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007634.hg.1 | NM_001002907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005536.hg.1 | ENST00000279783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005536.hg.1 | NM_001002907.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005536.hg.1 | TC11000451.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100007634.hg.1 | ENST00000279783.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007634.hg.1 | NM_001002907.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100153835.hg.1 | TC1100007634.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100153833.hg.1 | TC1100007634.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100153834.hg.1 | ENST00000279783.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100153834.hg.1 | NM_001002907.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100153834.hg.1 | ENST00000279783.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100153834.hg.1 | NM_001002907.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100153834.hg.1 | TC1100007634.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046401 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046401 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_16195-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 780129 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_16195 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1783643 | BC146844.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1783643 | NM_001002907.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1783643 | BC146844.1 | human_transcriptome |

OR8K1
Genomic Map