Assay Details
| Gene Symbol: OXCT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022120.1 | NP_071403.1 | 1 | 1041 | 166 | |
| GenBank mRNA | AB050193.2 | - | 1 | 967 | 166 | |
| AK093089.1 | - | 1 | 1015 | 166 | ||
| AK314586.1 | - | 1 | 948 | 166 | ||
| AY013700.1 | - | 1 | 1041 | 166 | ||
| BC030015.1 | - | 1 | 960 | 166 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002548.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20177327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153070 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20177317 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153080 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20177335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153071 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153074 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00153084 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P137896 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P85123 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P137896 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 30799 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 38170 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25425 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P137896 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P137896 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20177317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15051770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20177327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153069 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20177310 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153076 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153069 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153076 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15051770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20177317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20177310 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00153070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20177327 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P85123 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P137896 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00153075 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00153069 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00153072 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00153068 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00153067 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00153074 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15051740 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P137896 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P85123 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P124828 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P364829 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153086 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153075 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153080 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153068 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153072 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153084 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153079 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153074 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153078 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00153067 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15051740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7915259 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7900353 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7915258 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7915260 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7915257 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7900352 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 220256_PM_s_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 233301_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11758305_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11758305_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11757081_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11757081_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013805.hg.1 | ENST00000327582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013805.hg.1 | ENST00000327582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013805.hg.1 | ENST00000327582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013805.hg.1 | NM_022120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013805.hg.1 | NM_022120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013805.hg.1 | NM_022120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039619.hg.1 | ENST00000327582.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039619.hg.1 | ENST00000542949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039619.hg.1 | NM_022120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039619.hg.1 | TC01002548.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100013805.hg.1 | ENST00000327582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013805.hg.1 | ENST00000327582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013805.hg.1 | ENST00000327582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013805.hg.1 | NM_022120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013805.hg.1 | NM_022120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013805.hg.1 | NM_022120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203239.hg.1 | ENST00000327582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203239.hg.1 | NM_022120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203239.hg.1 | ENST00000327582.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203239.hg.1 | NM_022120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203239.hg.1 | TC0100013805.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 233301_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 233301_at | - | - |
| Human Genome U133 Plus 2.0 Array | 220256_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 220256_s_at | - | - |
| Human Genome U133A 2.0 Array | 220256_s_at | - | - |
| Human Genome U133A 2.0 Array | 220256_PM_s_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw551695 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220256_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 233301_at10 | - | - |
| Human Genome U95 Set | 57726_at | - | - |
| Human Genome U95 Set | 57726_PM_at | - | - |
| Human Genome U133 Set | 220256_s_at | - | - |
| Human Genome U133 Set | 220256_PM_s_at | - | - |
| Human Genome U133 Set | 233301_at | - | - |
| Human Genome U133 Set | 233301_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044039 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044039 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 152831 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 179599 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11545840-S | BC072388.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11545840-S | AY013700.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11545840-S | BC030015.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11545840-S | NM_022120.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11545840-S | AB050193.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_24641-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006290022 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006290022 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_24641 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_24641 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1653730 | AB050193.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1653730 | BC072388.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1653730 | AY013700.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1653730 | BC030015.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1653730 | NM_022120.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1653730 | AB050193.2 | human_transcriptome |

OXCT2
Genomic Map