Assay Details
| Gene Symbol: NBPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055895.1 | - | 3 | 2485 | 84 | |
| AK123260.1 | - | 3 | 2751 | 84 | ||
| BC110431.1 | - | 3 | 2485 | 84 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NBPF1
Genomic Map