Assay Details
| Gene Symbol: LDLRAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013693.2 | NP_001013715.2 | 1 - 2 | 270 | 57 | |
| XM_005245873.4 | XP_005245930.1 | 1 - 2 | 326 | 57 | ||
| GenBank mRNA | AK127275.1 | - | 1 - 2 | 270 | 57 | |
| BC146860.1 | - | 1 - 2 | 172 | 9056788 | 57 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000274.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01004218.hg.1 | ENST00000344642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004218.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004218.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004218.hg.1 | TC01000274.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004217.hg.1 | ENST00000344642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004217.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004217.hg.1 | TC01000274.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000484271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000484271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000484271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000484271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004219.hg.1 | ENST00000344642.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004219.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004219.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004219.hg.1 | TC01000274.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000484271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000484271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000484271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000484271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007254.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100150459.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150459.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150459.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100150459.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150459.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150459.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100150459.hg.1 | TC0100007254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100150457.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150457.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150457.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100150457.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150457.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150457.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100150457.hg.1 | TC0100007254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100150458.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150458.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150458.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100150458.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150458.hg.1 | ENST00000543870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150458.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100150458.hg.1 | TC0100007254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100150456.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150456.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100150456.hg.1 | ENST00000344642.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100150456.hg.1 | NM_001013693.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100150456.hg.1 | TC0100007254.hg.1 | Clariom D TC |

LDLRAD2
Genomic Map