Assay Details
| Gene Symbol: UPK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182684.2 | NP_872625.1 | 4 - 5 | 775 | 59 | |
| GenBank mRNA | AY233462.1 | - | 4 - 5 | 608 | 59 | |
| Gene Symbol: LOC105375355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006716140.3 | XP_006716203.1 | 4 - 5 | 853 | 59 | |
More Information
Set Membership: |
|

UPK3B
Genomic Map