Assay Details
| Gene Symbol: FBXO43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029860.3 | NP_001025031.2 | 1 - 2 | 823 | 83 | |
| XM_011516987.2 | XP_011515289.1 | 1 - 2 | 769 | 83 | ||
| XM_011516992.2 | XP_011515294.1 | 1 - 2 | 769 | 83 | ||
| XM_017013358.1 | XP_016868847.1 | 1 - 2 | 769 | 83 | ||
| GenBank mRNA | AK310518.1 | - | 1 - 2 | 393 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001475.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019136.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019136.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019136.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019136.hg.1 | TC08001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019138.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019138.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019138.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019138.hg.1 | TC08001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019139.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019139.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019139.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019139.hg.1 | TC08001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019141.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019141.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019141.hg.1 | TC08001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019142.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019142.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019142.hg.1 | TC08001475.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176589.hg.1 | TC0800011243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176591.hg.1 | TC0800011243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176593.hg.1 | TC0800011243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800176595.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176595.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176595.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176595.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176595.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176595.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176595.hg.1 | TC0800011243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800176596.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176596.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176596.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176596.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176596.hg.1 | TC0800011243.hg.1 | Clariom D TC |

FBXO43
Genomic Map